2-(3-Bromopropoxy)tetrahydropyran 98% stabilised
Leverantör: Thermo Fisher Scientific
Varning
Stabilization: Stabilised with potassium carbonate
399730050 399730010
ACRO399730050EA 3125 SEK
ACRO399730050 ACRO399730010
2-(3-Bromopropoxy)tetrahydropyran 98% stabilised
2-(3-Bromopropoxy)tetrahydropyran
Formel: C₈H₁₅BrO₂
Kokpunkt: 64,5 °C (.8 mmHg)
Densitet: 1,314 g/cm³
Flampunkt: 106 °C
Storage Temperature: Frys
MDL Number: MFCD00058593
CAS nummer: 33821-94-2
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4770 to 1.4790 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer K2CO3