Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2) 97%
Leverantör: Thermo Scientific
Synonyms: Cytidine-5'-triphosphate disodium, Sodium ((2R,3S,4R,5R)-5-(4-amino-2-oxopyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl dihydrogentriphosphate
J62238.03 J62238.04 J62238.ME
J62238.03EA 8900 SEK
J62238.03 J62238.04 J62238.ME
Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2) 97%
Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2)
Purity: 97+% CAS No.: 36051-68-0 Molecular Formula: C9H14N3Na2O14P3 Formula Weight: 527.12 Flash Point: 113°(235°F) Physical Form: Powder MDL No.: MFCD00078193
Formel: C₉H₁₄N₃Na₂O₁₄P₃
MW: 527,12 g/mol
Flampunkt: 113 °C (235 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00078193
CAS nummer: 36051-68-0
EINECS: 252-849-3