2-Chloroadenosine
Leverantör: Thermo Fisher Scientific
Synonyms:
(2R,3R,4S,5R)-2-(6-Amino-2-chloro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol
J63810.MD
J63810.03
J63810.MDEA
2925
SEK
J63810.MD
J63810.03
2-Chloroadenosine
2-Chloroadenosine
CAS No.: 146-77-0
Molecular Formula: C10H12ClN5O4
Formula Weight: 301.69
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Beilstein Reference: 43957
MDL No.: MFCD00005734
A selective A1-adenosine receptor agonist. Induces apoptosis
Formel:
C₁₀H₁₂ClN₅O₄ MW: 301.69 g/mol |
MDL Number:
MFCD00005734 CAS nummer: 146-77-0 EINECS: 205-678-3 |