Ethyl (ethoxymethylene)acetate ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 3-Ethoxyacrylic acid ethyl ester, Ethyl 3-Ethoxyacrylate, Ethyl 3-Ethoxy-2-Propenoate, Ethyl 3-ethoxyacrylate
H59187.09 H59187.18
H59187.09EA 2100 SEK
H59187.09 H59187.18
Ethyl (ethoxymethylene)acetate ≥98%
Ethyl (ethoxymethylene)acetate
Formel: C₇H₁₂O₃
Kokpunkt: 195…196 °C
Densitet: 0,998
Flampunkt: 76 °C (169 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00009863
CAS nummer: 1001-26-9
UN: 3334
ADR: 9,