Zinkoxid, NanoArc® ZN-0605
Leverantör: Thermo Fisher Scientific
Varning
45849.22 45849.36
45849.22EA 725 SEK
45849.22 45849.36
Zinkoxid, NanoArc® ZN-0605
Zinkoxid
CAS No.: 1314-13-2 Molecular Formula: OZn Formula Weight: 81.37 Physical Form: 40-100nm APS Powder Appearance: White Odour: Odourless Merck Reference: 14,10147 UN No.: 3077 MDL No.: MFCD00011300
Formel: ZnO
Smältpunkt: 1975 °C
Densitet: 5,61 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011300
CAS nummer: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9,III
REACH: 01-2119463881-32
Merck Index: 14,10147