Magnesiumklorid, ren
Leverantör: Thermo Fisher Scientific
223211000 223210010 223210025 223210250
ACRO223211000EA 810 SEK
ACRO223211000 ACRO223210010 ACRO223210025 ACRO223210250
Magnesiumklorid, ren
Magnesiumklorid
Formel: MgCl₂
MW: 95.21 g/mol
Kokpunkt: 1412 °C (760 mmHg)
Smältpunkt: 708 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011106
CAS nummer: 7786-30-3
EINECS: 232-094-6
Merck Index: 13,05684
Order Now
Specifikation testresultat
Appearance White Powder (May contain grey to black particles)
Titration Complexometric ≥98.0 % (on dry substance)
Water ≤3 % (K.F.)