3-Bromo-5-formylpyridine ≥98.0%
Leverantör: TCI
Varning
Synonyms: 3-Bromopyridine-5-carboxaldehyde, 5-Bromopyridine-3-carboxaldehyde, 5-Bromonicotinaldehyde, 5-Bromo-3-pyridinecarboxaldehyde
B3490-1G B3490-5G
TCIAB3490-1GEA 375 SEK
TCIAB3490-1G TCIAB3490-5G
3-Bromo-5-formylpyridine ≥98.0%
3-Bromo-5-formylpyridine
Formel: C₆H₄BrNO
MW: 186.01 g/mol
MDL Number: MFCD03265758
CAS nummer: 113118-81-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4544e913b0397002377c9c779cfe36d831ee33b4&vid=4544e913b0397002377c9c779cfe36d831ee33b4&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 96.0 to 100.0 deg-C