α,3,5-Tribromotoluene ≥95.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
3,5-Dibromobenzyl bromide, 1,3-Dibromo-5-(bromomethyl)benzene, alpha,3,5-Tribromotoluene
D4020-5G
D4020-25G
TCIAD4020-5GEA
630
SEK
TCIAD4020-5G
TCIAD4020-25G
α,3,5-Tribromotoluene ≥95.0%
α,3,5-Tribromotoluene
Formel:
C₇H₅Br₃ MW: 328,83 g/mol Kokpunkt: 169°C Smältpunkt: 93…95 °C |
MDL Number:
MFCD00052415 CAS nummer: 56908-88-4 UN: 3261 ADR: 8,III |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 92.0 to 96.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...