1-Aminocyclopropanecarboxylic acid 98%
Leverantör: Thermo Scientific
Varning
Synonyms: 1-Amino-1-cyclopropanecarboxylic acid
J65118.MD J65118.03
J65118.MDEA 1750 SEK
J65118.MD J65118.03
1-Aminocyclopropanecarboxylic acid 98%
1-Aminocyclopropanecarboxylic acid
Formel: C₄H₇NO₂
MW: 101,11 g/mol
Smältpunkt: 229…231 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00009944
CAS nummer: 22059-21-8