L-Threonine Cell Culture
Leverantör: Thermo Fisher Scientific
Synonyms: (2S,3R)-(-)-Threonine, H-Thr-OH, (2S,3R)-2-Amino-3-hydroxybutyric acid
J63709.30 J63709.A1
J63709.30EA 1650 SEK
J63709.30 J63709.A1
L-Threonine Cell Culture
L-Threonine
CAS No.: 72-19-5 Molecular Formula: C4H9NO3 Formula Weight: 119.12 Physical Form: Powder Appearance: White Melting Point: 256° dec. Beilstein Reference: 1721646 Merck Reference: 14,9380 MDL No.: MFCD00064270
Formel: C₄H₉NO₃
MW: 119.12 g/mol
Smältpunkt: 256 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00064270
CAS nummer: 72-19-5
EINECS: 200-774-1
Merck Index: 14,9380