Magnesiumklorid hexahydrat cell culture reagent
Leverantör: Thermo Fisher Scientific
J62575.30 J62575.36 J62575.A1
J62575.30EA 1425 SEK
J62575.30 J62575.36 J62575.A1
Magnesiumklorid hexahydrat cell culture reagent
Magnesiumklorid hexahydrat
CAS No.: 7791-18-6 Molecular Formula: MgCl2.6H2O Formula Weight: 203.30 (95.22anhy) Physical Form: Powder Appearance: White Odour: Odourless Merck Reference: 14,5662 MDL No.: MFCD00149781
Formel: MgCl₂·6H₂O
Kokpunkt: 1412 °C (1013 hPa)
Smältpunkt: 117 °C
Densitet: 1,57 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00149781
CAS nummer: 7791-18-6
EINECS: 232-094-6
Merck Index: 14,5662