Olja av cederträ

Leverantör: TCI
Varning

Synonyms: Cedar wood oil, Cedar Oil, Cedar Leaf Oil

C0626-500ML C0626-25ML C0626-100ML
TCIAC0626-500MLEA 3825 SEK
TCIAC0626-500ML TCIAC0626-25ML TCIAC0626-100ML
Olja av cederträ
Olja av cederträ
Storage Temperature: Omgivande
MDL Number: MFCD00132766
CAS nummer: 8000-27-9

Order Now

Specifikation testresultat

Appearance White to Yellow to Orange clear liquid
Acid Value max. 20.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR