Natriumalginat, high viscosity
Leverantör: Thermo Fisher Scientific
CAS No.: 9005-38-3
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White to off-white
Merck Reference: 14,241
MDL No.: MFCD00081310
A gelling polysaccharide extracted from giant brown seaweed, useful in viscosity studies
Storage Temperature:
Omgivande |
MDL Number:
MFCD00081310 CAS nummer: 9005-38-3 Merck Index: 14,241 |