DL-Cystine
Leverantör: Thermo Fisher Scientific
Synonyms: (H-DL-Cys-OH)2
J63564.06 J63564.14 J63564.22
J63564.06EA 1100 SEK
J63564.06 J63564.14 J63564.22
DL-Cystine
DL-Cystine
CAS No.: 923-32-0 Molecular Formula: C6H14N2O4S2 Formula Weight: 240.30 Physical Form: Powder Appearance: White Melting Point: 227° dec. Beilstein Reference: 1728095 Merck Reference: 14,2782 MDL No.: MFCD00084652
Formel: C₆H₁₂N₂O₄S₂
MW: 240.3 g/mol
Kokpunkt: 429
Smältpunkt: 227 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00084652
CAS nummer: 923-32-0
EINECS: 213-094-5
Merck Index: 13,02811