Magnesiumsulfat heptahydrat cell culture reagent
Leverantör: Thermo Fisher Scientific
Synonyms: sulphuric acid magnesium salt heptahydrate, Epsom salt, Bitter salt
J61839.36 J61839.A1
J61839.36EA 1700 SEK
J61839.36 J61839.A1
Magnesiumsulfat heptahydrat cell culture reagent
Magnesiumsulfat heptahydrat
CAS No.: 10034-99-8 Molecular Formula: MgO4S.7H2O Formula Weight: 246.48 (120.37anhy) Physical Form: Powder Density: 1.68 Appearance: White Odour: Odourless Merck Reference: 14,5691 MDL No.: MFCD00149785
Formel: MgSO₄·7H₂O
MW: 246.48 g/mol
Smältpunkt: 1124
Densitet: 1.68
Storage Temperature: Omgivande
MDL Number: MFCD00149785
CAS nummer: 10034-99-8
EINECS: 231-298-2
Merck Index: 14,5691