L (+)-glutamin cell culture reagent
Leverantör: Thermo Scientific
Synonyms: Gln, L-Glutamine, H-Gln-OH, (S)-(+)-Glutamine, (S)-2,5-Diamino-5-oxopentanoic acid, Levoglutamide, Q
J60573.14 J60573.22 J60573.A1
J60573.14EA 765 SEK
J60573.14 J60573.22 J60573.A1
L (+)-glutamin cell culture reagent
L (+)-glutamin
CAS No.: 56-85-9 Molecular Formula: C5H10N2O3 Formula Weight: 146.14 Physical Form: Crystalline powder Appearance: White pH: 5.0-6.0 Merck Reference: 14,4471 MDL No.: MFCD00008044
Formel: C₅H₁₀N₂O₃
MW: 146,15 g/mol
Kokpunkt: 355 °C (1013 hPa)
Smältpunkt: 185…186 °C
Densitet: 1,5326 g/cm³ (130 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00008044
CAS nummer: 56-85-9
EINECS: 200-292-1
Merck Index: 14,4471
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb7a812ec5d2c73a797fc464926df9eef12c5c9a&vid=f0456e1fafdcae9257ac25b71cb204be7448e497&prid=web&clid=SAPDAM