D (+)-Sackaros för molekylärbiologi
Leverantör: Thermo Fisher Scientific
CAS No.: 57-50-1
Molecular Formula: C12H22O11
Formula Weight: 342.30
Physical Form: Crystalline solid
Density: 1.58
Appearance: White
Odour: Odourless
Melting Point: 185-187°
Solubility: Soluble in water
Beilstein Reference: 90825
MDL No.: MFCD00006626
For protein purification by sucrose density gradients
Formel:
C₁₂H₂₂O₁₁ Smältpunkt: 185…187 °C Densitet: 1,58 Storage Temperature: Omgivande |
MDL Number:
MFCD00006626 CAS nummer: 57-50-1 EINECS: 200-334-9 Merck Index: 13,08966 |