Tris-(hydroximetyl) aminometan 99.5% för elektrofores
Leverantör: Thermo Fisher Scientific
Varning
J61016.22 J61016.36
J61016.22EA 1075 SEK
J61016.22 J61016.36
Tris-(hydroximetyl) aminometan 99.5% för elektrofores
Tris-(hydroximetyl) aminometan
Purity: 99.5% CAS No.: 77-86-1 Molecular Formula: C4H11NO3 Formula Weight: 121.14 Physical Form: Crystalline powder Appearance: White Solubility: Freely soluble in water Beilstein Reference: 741883 Merck Reference: 14,9772 MDL No.: MFCD00004679
Formel: NH₂C(CH₂OH)₃
Kokpunkt: 357 °C (1013 hPa)
Smältpunkt: 169…172 °C
Densitet: 1,328 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00004679
CAS nummer: 77-86-1
EINECS: 201-064-4
Merck Index: 14,9772
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM