Dibenzyl dicarbonate ≥97.0%

Leverantör: TCI
Varning

Synonyms: Dibenzyl pyrocarbonate

P1281-25G P1281-5G
TCIAP1281-25GEA 4425 SEK
TCIAP1281-25G TCIAP1281-5G
Dibenzyl dicarbonate ≥97.0%
Dibenzyl dicarbonate
Formel: C₁₆H₁₄O₅
MW: 286.28 g/mol
Storage Temperature: Frys
MDL Number: MFCD00043124
CAS nummer: 31139-36-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=044c13831c21cfddbdbccf1e01a9ee643ad4a2f9&vid=044c13831c21cfddbdbccf1e01a9ee643ad4a2f9&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Morpholine Method) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR