4-Bromobenzylamine hydrochloride 98%
Leverantör: Thermo Scientific
Varning
Synonyms: (4-Bromophenyl)methanamine hydrochloride
A10979.09 A10979.18
A10979.09EA 3650 SEK
A10979.09 A10979.18
4-Bromobenzylamine hydrochloride 98%
4-Bromobenzylamine hydrochloride
Purity: 98% CAS No.: 26177-44-6 Molecular Formula: C7H8BrN.HCl Formula Weight: 222.52 Melting Point: 272-274° Beilstein Reference: 3693458
Formel: C₇H₉BrClN
MW: 222,52 g/mol
Smältpunkt: 272…274 °C
Storage Temperature: Omgivande
MDL Number: MFCD00012860
CAS nummer: 26177-44-6
EINECS: 247-503-3