2-Bromobenzylamine hydrochloride ≥98.0%

Leverantör: TCI EUROPE
Varning

B1836-25G B1836-5G
TCIAB1836-25GEA 2650 SEK
TCIAB1836-25G TCIAB1836-5G
2-Bromobenzylamine hydrochloride ≥98.0%
2-Bromobenzylamine hydrochloride
Formel: C₇H₉BrClN
MW: 222,52 g/mol
MDL Number: MFCD00012853
CAS nummer: 5465-63-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e3f36f9fdf7a96fa98c013a8234cf8283deb0dbd&vid=e3f36f9fdf7a96fa98c013a8234cf8283deb0dbd&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR