1,3-Dibromo-3-methylbutane ≥98%
Leverantör: Thermo Scientific
Varning
B20468.06 B20468.14 B20468.22
B20468.06EA 795 SEK
B20468.06 B20468.14 B20468.22
1,3-Dibromo-3-methylbutane ≥98%
1,3-Dibromo-3-methylbutane
Purity: 98% CAS No.: 24443-15-0 Molecular Formula: (CH3)2CBrCH2CH2Br Formula Weight: 229.95 Flash Point: 110°(230°F)
Formel: C₅H₁₀Br₂
MW: 229,95 g/mol
Densitet: 1,69
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00060773
CAS nummer: 24443-15-0