Dibenzyl carbonate ≥98.0%

Leverantör: TCI
Varning

C1600-1G C1600-5G
TCIAC1600-1GEA 415 SEK
TCIAC1600-1G TCIAC1600-5G
Dibenzyl carbonate ≥98.0%
Dibenzyl carbonate
Formel: C₁₅H₁₄O₃
MDL Number: MFCD00014436
CAS nummer: 3459-92-5
EINECS: 222-401-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3397054beb9086a67d0a2b989265a1d555cfa8ef&vid=3397054beb9086a67d0a2b989265a1d555cfa8ef&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR