Tetrahydro-4H-thiopyran-4-one ≥98.0%

Leverantör: TCI EUROPE

Synonyms: Tetrahydrothiopyran-4-one

O0112-25G O0112-5G
TCIAO0112-25GEA 1650 SEK
TCIAO0112-25G TCIAO0112-5G
Tetrahydro-4H-thiopyran-4-one ≥98.0%
Tetrahydro-4H-thiopyran-4-one
Formel: C₅H₈OS
MW: 116,18 g/mol
MDL Number: MFCD00006660
CAS nummer: 1072-72-6
EINECS: 214-015-7

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 61.0 to 65.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR