Samarium(III) perchlorate 50% (w/w) i vattenlösning, Reagent Grade
Leverantör: Thermo Fisher Scientific
Fara
40567.09 40567.18
40567.09EA 2250 SEK
40567.09 40567.18
Samarium(III) perchlorate 50% (w/w) i vattenlösning, Reagent Grade
Samarium(III) perchlorate
CAS No.: 13569-60-3 Molecular Formula: Sm(ClO4)3.6H2O Formula Weight: 556.80 (448.70anhy) Physical Form: Liquid Odour: Odourless UN No.: 3211 MDL No.: MFCD00049769
Formel: Cl₃O₁₂Sm
MW: 448.698 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00049769
CAS nummer: 13569-60-3
EINECS: 236-986-6
UN: 3211
ADR: 5.1 II