4'-Fluoro-3'-methoxyacetophenone ≥98%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 1-(4-Fluoro-3-methoxyphenyl)ethanone
L19894.03 L19894.MD
L19894.03EA 4950 SEK
L19894.03 L19894.MD
4'-Fluoro-3'-methoxyacetophenone ≥98%
4'-Fluoro-3'-methoxyacetophenone
Purity: 98% CAS No.: 64287-19-0 Molecular Formula: C9H9FO2 Formula Weight: 168.17 Melting Point: 51-53° Beilstein Reference: 4381319
Formel: C₉H₉FO₂
MW: 168.17 g/mol
Smältpunkt: 51…53 °C
Storage Temperature: Omgivande
MDL Number: MFCD00272135
CAS nummer: 64287-19-0