4-Fluoro-3-methylphenylboronic acid (contains varying amounts of Anhydride)

Leverantör: TCI EUROPE
Varning

Synonyms: 3-Methyl-4-fluorophenylboronic acid, 4-Fluoro-3-methylbenzeneboronic acid

F0697-25G F0697-5G
TCIAF0697-25GEA 2700 SEK
TCIAF0697-25G TCIAF0697-5G
4-Fluoro-3-methylphenylboronic acid (contains varying amounts of Anhydride)
4-Fluoro-3-methylphenylboronic acid
Formel: C₇H₈BFO₂
MW: 153,95 g/mol
MDL Number: MFCD01863527
CAS nummer: 139911-27-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97281b30414a051aa2703b1590bec369556baa30&vid=97281b30414a051aa2703b1590bec369556baa30&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 97.0 to 113.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR