4-(Hexyloxy)phenylboronic acid (contains varying amounts of Anhydride)
Leverantör: TCI
|
Varning
|
Synonyms:
B-[4-(Hexyloxy)phenyl]boronic acid, 4-n-Hexyloxybenzeneboronic acid
H1357-5G
H1357-1G
TCIAH1357-5GEA
1275
SEK
TCIAH1357-5G
TCIAH1357-1G
4-(Hexyloxy)phenylboronic acid (contains varying amounts of Anhydride)
4-(Hexyloxy)phenylboronic acid
Formel:
C₁₂H₁₉BO₃ Smältpunkt: 85°C |
MDL Number:
MFCD04039027 CAS nummer: 121219-08-7 |
|
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 109.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...