Sodium-1-undecanesulphonate ≥98.0% för jonparkromatografi

Leverantör: TCI

I0349-25G I0349-5G
TCIAI0349-25GEA 2250 SEK
TCIAI0349-25G TCIAI0349-5G
Sodium-1-undecanesulphonate ≥98.0% för jonparkromatografi
Formel: C₁₂H₂₅NaO₃S
MDL Number: MFCD00053627
CAS nummer: 5838-34-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d26cd2a4605d7accef8cd68136c4bf0af2f51eb&vid=78e4ba6e3f362327d5b32b0f2760a5ae2dca43a8&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White powder to crystal
Purity (Ion exchange titration) min. 98.0 %
Absorbance (210nm) max. 1.000
Absorbance (230nm) max. 0.300
Absorbance (254nm) max. 0.130


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR