Sodium-1-octadecanesulphonate 90%
Leverantör: Thermo Fisher Scientific
384910010 384910050 384910100
ACRO384910010EA 470 SEK
ACRO384910010 ACRO384910050 ACRO384910100
Sodium-1-octadecanesulphonate 90%
Sodium-1-octadecanesulphonate
Formel: C₁₈H₃₇NaO₃S
Smältpunkt: 163…165 °C
Storage Temperature: Omgivande
MDL Number: MFCD00051526
CAS nummer: 13893-34-0
EINECS: 237-660-6
Order Now
Specifikation testresultat
Appearance Off-white Powder
Infrared spectrum Conforms
Loss on drying ≤5 % (1 g, 105°C, 1 h, 760 mmHg)
Elemental Analysis ≥88 % (C, H)