3,5-Dimethoxybenzyl bromide ≥97.0%
Leverantör: TCI EUROPE
Fara
Synonyms: 1-(Bromomethyl)-3,5-dimethoxybenzene
D2657-25G D2657-5G
TCIAD2657-25GEA 2950 SEK
TCIAD2657-25G TCIAD2657-5G
3,5-Dimethoxybenzyl bromide ≥97.0%
3,5-Dimethoxybenzyl bromide
Formel: C₉H₁₁BrO₂
MW: 231,0864 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD01321368
CAS nummer: 877-88-3
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=67d41cffa15e2b95396b6fa9b81f66024e23839d&vid=67d41cffa15e2b95396b6fa9b81f66024e23839d&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 71.0 to 74.0 deg-C
Solubility in Methanol almost transparency