3-Pyridinesulphonic acid 98%
Leverantör: Thermo Scientific
Fara
Synonyms: Pyridine-3-sulphonic acid
131921000 131925000
ACRO131921000EA 2975 SEK
ACRO131921000 ACRO131925000
3-Pyridinesulphonic acid 98%
3-Pyridinesulphonic acid
Formel: C₅H₅NO₃S
MW: 159,16 g/mol
Smältpunkt: >300 °C
Flampunkt: >66 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006381
CAS nummer: 636-73-7
EINECS: 211-265-9
UN: 2585
ADR: 8,III
Order Now
Specifikation testresultat
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Solubility (5 % in water) Clear colorless
Titration with NaOH ≥97.5 %
Elemental Analysis 9.92 to 10.43 % N
Specific optical rotation +5° to +8° (20°C, 589 nm) (c=2, methanol)