Silicon nitride amorphous ≥98.5%, nanopowder
Leverantör: Thermo Scientific
Synonyms: Silicon(IV) nitride
45611.06 45611.14 45611.22
45611.06EA 1350 SEK
45611.06 45611.14 45611.22
Silicon nitride amorphous ≥98.5%, nanopowder
Silicon nitride
Purity: 98.5+% CAS No.: 12033-89-5 Molecular Formula: Si3N4 Formula Weight: 140.28 Physical Form: 15-30nm APS Powder, S.A. 103-123m2/g Density: 3.2 Appearance: Pale grey Odour: Odourless Melting Point: 1900° subl. Merck Reference: 14,8497 MDL No.: MFCD00011230
Formel: N₄Si₃
MW: 140,28 g/mol
Smältpunkt: 1900 °C (subl.)
Densitet: 3,2
Storage Temperature: Omgivande
MDL Number: MFCD00011230
CAS nummer: 12033-89-5
EINECS: 234-796-8
Merck Index: 14,8497