Silicon nitride α-phase ≥99.85% 90+% α-phase, Electronic Grade
Leverantör: Thermo Scientific
Synonyms: Silicon(IV) nitride
49113.09 49113.18
49113.09EA 765 SEK
49113.09 49113.18
Silicon nitride α-phase ≥99.85% 90+% α-phase, Electronic Grade
Silicon nitride
Purity: 99.85% CAS No.: 12033-89-5 Molecular Formula: N4Si3 Formula Weight: 140.28 Physical Form: -325 Mesh Powder Density: 3.2 Appearance: Pale gray Odour: Odourless Melting Point: 1900° subl. Merck Reference: 14,8497 MDL No.: MFCD00011230
Formel: N₄Si₃
MW: 140,28 g/mol
Smältpunkt: 1900 °C (subl.)
Densitet: 3,2
Storage Temperature: Omgivande
MDL Number: MFCD00011230
CAS nummer: 12033-89-5
EINECS: 234-796-8
Merck Index: 14,8497