Calcium nitride ≥99%
Leverantör: Thermo Scientific
Fara
13124.06 13124.18 13124.30
13124.06EA 2650 SEK
13124.06 13124.18 13124.30
Calcium nitride ≥99%
Calcium nitride
Purity: 99% CAS No.: 12013-82-0 Molecular Formula: Ca3N2 Formula Weight: 148.25 Physical Form: 12mm & Down Pieces Density: 2.63 Melting Point: 1195° UN No.: 3178 MDL No.: MFCD00015985
Formel: Ca₃N₂
MW: 148,25 g/mol
Smältpunkt: 1195 °C
Densitet: 2,63
Storage Temperature: Omgivande
MDL Number: MFCD00015985
CAS nummer: 12013-82-0
EINECS: 234-592-9
UN: 3178
ADR: 4.1,II