Potassium selenocyanate 99%
Leverantör: Thermo Fisher Scientific
Fara
201980100 201980500 201982500
ACRO201980100EA 1700 SEK
ACRO201980100 ACRO201980500 ACRO201982500
Potassium selenocyanate 99%
Potassium selenocyanate
Formel: CKNSe
Smältpunkt: 152…158 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011366
CAS nummer: 3425-46-5
EINECS: 222-320-1
UN: 3283
ADR: 6.1,II
Order Now
Specifikation testresultat
Appearance White to beige Crystalline powder or needles
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %