cis-4-Cyclohexene-1,2-dicarboxylic acid ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: (1R,2S)-rel-Cyclohex-4-ene-1,2-dicarboxylic acid
L09392.14 L09392.22
L09392.14EA 970 SEK
L09392.14 L09392.22
cis-4-Cyclohexene-1,2-dicarboxylic acid ≥98%
cis-4-Cyclohexene-1,2-dicarboxylic acid
Purity: 98% CAS No.: 2305-26-2 Molecular Formula: C8H10O4 Formula Weight: 170.17 Melting Point: 165-170° Beilstein Reference: 2050419
Formel: C₈H₁₀O₄
MW: 170,17 g/mol
Smältpunkt: 165…170 °C
Storage Temperature: Omgivande
MDL Number: MFCD00070482
CAS nummer: 2305-26-2
EINECS: 218-974-2