2,4-Dichloro-6-methylbenzylamine ≥98%
Leverantör: Thermo Fisher Scientific
|
Fara
|
L16108.03
L16108.06
L16108.03EA
2575
SEK
L16108.03
L16108.06
2,4-Dichloro-6-methylbenzylamine ≥98%
2,4-Dichloro-6-methylbenzylamine
Purity: 98+%
CAS No.: 150517-76-3
Molecular Formula: C8H9Cl2N
Formula Weight: 190.07
Melting Point: 52-56°
Beilstein Reference: 8615227
UN No.: 3259
MDL No.: MFCD00052913
Formel:
C₈H₉Cl₂N Smältpunkt: 52…56 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00052913 CAS nummer: 150517-76-3 ADR: 8,III |