2-Bromo-3'-(trifluoromethyl)acetophenone ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 3-(Trifluoromethyl)phenacyl bromide, 2-Bromo-1-(3-(trifluoromethyl)phenyl)ethanone
H31526.03 H31526.06 H31526.09
H31526.03EA 2250 SEK
H31526.03 H31526.06 H31526.09
2-Bromo-3'-(trifluoromethyl)acetophenone ≥98%
2-Bromo-3'-(trifluoromethyl)acetophenone
Purity: 98% CAS No.: 2003-10-3 Molecular Formula: C9H6BrF3O Formula Weight: 267.04 Physical Form: Fused solid Appearance: Pale yellow Melting Point: 20-23° Boiling Point: 69-72° MDL No.: MFCD03094283
Formel: C₉H₆BrF₃O
MW: 267,04 g/mol
Kokpunkt: 69…72 °C
Smältpunkt: 20…23 °C
Storage Temperature: Omgivande
MDL Number: MFCD03094283
CAS nummer: 2003-10-3