2-Bromo-2'-(trifluoromethyl)acetophenone ≥97%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-(Trifluoromethyl)phenacyl bromide, 2-Bromo-1-[2-(trifluoromethyl)phenyl]-1-ethanone
H32032.03 H32032.06
H32032.03EA 1800 SEK
H32032.03 H32032.06
2-Bromo-2'-(trifluoromethyl)acetophenone ≥97%
2-Bromo-2'-(trifluoromethyl)acetophenone
Purity: 97% CAS No.: 54109-16-9 Molecular Formula: C9H6BrF3O Formula Weight: 267.04 Melting Point: 24-32° MDL No.: MFCD03094304
Formel: C₉H₆BrF₃O
MW: 267,04 g/mol
Smältpunkt: 24…32 °C
Storage Temperature: Omgivande
MDL Number: MFCD03094304
CAS nummer: 54109-16-9
UN: 3265
ADR: 8,III