4-Hydroxy-3-methoxybenzylamine hydrochloride 98%
Leverantör: Thermo Scientific
Varning
Synonyms: Vanillylamine HCl, 4-(Aminomethyl)-2-methoxyphenol hydrochloride
B20637.06 B20637.14
B20637.06EA 1875 SEK
B20637.06 B20637.14
4-Hydroxy-3-methoxybenzylamine hydrochloride 98%
4-Hydroxy-3-methoxybenzylamine hydrochloride
Purity: 98% CAS No.: 7149-10-2 Molecular Formula: C8H11NO2.HCl Formula Weight: 189.64 Melting Point: ca 220° dec. Beilstein Reference: 3915418
Formel: C₈H₁₂ClNO₂
MW: 189,64 g/mol
Smältpunkt: ∼ 220 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00012864
CAS nummer: 7149-10-2
EINECS: 230-468-3