4-Hydroxy-3-methoxybenzylamine hydrochloride 98%
Leverantör: Thermo Scientific
Varning
Synonyms: Vanillylamine HCl, 4-(Aminomethyl)-2-methoxyphenol hydrochloride
121370010 121370050
ACRO121370010EA 625 SEK
ACRO121370010 ACRO121370050
4-Hydroxy-3-methoxybenzylamine hydrochloride 98%
4-Hydroxy-3-methoxybenzylamine hydrochloride
Formel: C₈H₁₂ClNO₂
MW: 189,64 g/mol
Smältpunkt: 219…221 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00012864
CAS nummer: 7149-10-2
EINECS: 230-468-3
Order Now
Specifikation testresultat
Appearance Off-white Powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %
GC ≥95 %