5'-Bromo-2'-hydroxy-3'-nitroacetophenone ≥97%
Leverantör: Thermo Fisher Scientific
Varning
H27331.14 H27331.06
H27331.14EA 2675 SEK
H27331.14 H27331.06
5'-Bromo-2'-hydroxy-3'-nitroacetophenone ≥97%
5'-Bromo-2'-hydroxy-3'-nitroacetophenone
Purity: 97% CAS No.: 70978-54-0 Molecular Formula: C8H6BrNO4 Formula Weight: 260.05 Melting Point: 129-132° MDL No.: MFCD01631134
Formel: C₈H₆BrNO₄
Smältpunkt: 129…132 °C
Storage Temperature: Omgivande
MDL Number: MFCD01631134
CAS nummer: 70978-54-0