N-Fluorobenzenesulphonimide 97%
Leverantör: Thermo Scientific
Varning
311900050 311900250
ACRO311900050EA 1275 SEK
ACRO311900050 ACRO311900250
N-Fluorobenzenesulphonimide 97%
N-Fluorobenzenesulphonimide
Formel: C₁₂H₁₀FNO₄S₂
MW: 315,34 g/mol
Smältpunkt: 110…115 °C
Storage Temperature: Omgivande
MDL Number: MFCD00144885
CAS nummer: 133745-75-2
Order Now
Specifikation testresultat
Appearance Off-white to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 110 to 117 °C
Titration with Na2S2O3 ≥96.0 %