(S)-(-)-N-Benzyl-α-methylbenzylamine ≥99%, ee 99%, ChiPros®
Leverantör: Thermo Scientific
Varning
Synonyms: S-(-)-N-Benzyl-1-phenylethylamine, (S)-(-)-N-Benzyl-1-phenylethylamine
L19299.06 L19299.14
L19299.06EA 935 SEK
L19299.06 L19299.14
(S)-(-)-N-Benzyl-α-methylbenzylamine ≥99%, ee 99%, ChiPros®
(S)-(-)-N-Benzyl-α-methylbenzylamine
Purity: 99% CAS No.: 17480-69-2 Molecular Formula: C15H17N Formula Weight: 211.31 Flash Point: 110°(230°F) Refractive Index: 1.5640 Boiling Point: 171°/15mm Beilstein Reference: 3650264 Optical Rotation: -39° (neat) MDL No.: MFCD00066325
Formel: C₁₅H₁₇N
MW: 211,31 g/mol
Kokpunkt: 171 °C (15 mmHg)
Densitet: 1,010
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00066325
CAS nummer: 17480-69-2