Allyl methyl carbonate ≥98.0%

Leverantör: TCI EUROPE
Varning

C1315-25ML C1315-5ML
TCIAC1315-25MLEA 2575 SEK
TCIAC1315-25ML TCIAC1315-5ML
Allyl methyl carbonate ≥98.0%
Allyl methyl carbonate
Formel: C₅H₈O₃
MW: 116,1152 g/mol
Kokpunkt: 131°C
Flampunkt: 33°C
MDL Number: MFCD00134545
CAS nummer: 35466-83-2
EINECS: 468-750-5
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6970f90464f55cb41b05744a0ba88cc183712ec5&vid=6970f90464f55cb41b05744a0ba88cc183712ec5&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR