2-Chloroethyl ethyl sulphide ≥98.0%

Leverantör: TCI EUROPE
Fara

C0938-25ML C0938-5ML
TCIAC0938-25MLEA 2375 SEK
TCIAC0938-25ML TCIAC0938-5ML
2-Chloroethyl ethyl sulphide ≥98.0%
2-Chloroethyl ethyl sulphide
Formel: C₄H₉ClS
Kokpunkt: 157°C
Flampunkt: 52°C
Storage Temperature: Kylskåp
MDL Number: MFCD00000979
CAS nummer: 693-07-2
EINECS: 211-742-1
UN: 1992
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f14518c8056a5b7e5d31cebc436eec2a504a9bc1&vid=f14518c8056a5b7e5d31cebc436eec2a504a9bc1&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR