5-(3-Hydroxyphenyl)-1H-tetrazole ≥97%
Leverantör: Thermo Fisher Scientific
Varning
H27529.MD H27529.06 H27529.03
H27529.MDEA 1250 SEK
H27529.MD H27529.06 H27529.03
5-(3-Hydroxyphenyl)-1H-tetrazole ≥97%
5-(3-Hydroxyphenyl)-1H-tetrazole
Purity: 97% CAS No.: 96859-34-6 Molecular Formula: C7H6N4O Formula Weight: 162.16 Melting Point: ca 220° dec. MDL No.: MFCD06797247
Formel: C₇H₆N₄O
Smältpunkt: ∼ 220 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD06797247
CAS nummer: 96859-34-6