Benzothiazol-2-ol
Leverantör: Thermo Scientific
Synonyms:
2-Hydroxybenzothiazole
AC40197DA
AC40197EA
MAYBAC40197DAEA
0
SEK
MAYBAC40197DA
MAYBAC40197EA
Benzothiazol-2-ol
Benzothiazol-2-ol
Formel:
C₇H₅NOS MW: 151.18 g/mol |
MDL Number:
MFCD00022868 CAS nummer: 934-34-9 EINECS: 213-281-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...