Furan-2,5-dicarboxylic acid ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
2,5-Furandicarboxylic acid
F0710-25G
F0710-5G
TCIAF0710-25GEA
3275
SEK
TCIAF0710-25G
TCIAF0710-5G
Furan-2,5-dicarboxylic acid ≥98.0% (by HPLC, titration analysis)
Furan-2,5-dicarboxylic acid
Formel:
C₆H₄O₅ MW: 156,09 g/mol Smältpunkt: 320°C |
MDL Number:
MFCD00016582 CAS nummer: 3238-40-2 EINECS: 221-800-8 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...